Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • In Stock (3)
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "portable-balances"
https://ru.vwr.com/stibo/smallweb/std.lang.all/40/64/18374064.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"18"
  List View Grid View Easy View (new)
Sort by:
Catalog Number: (611-3131)
Supplier:  VWR Collection
Description:   A complete portable system for density measurements, comprising of a portable balance plus a density kit for solids and liquids with the required acce...
UOM:  1 * 1 KIT
Supplier:  VWR Collection
Description:   A unique product that combines the simplicity of a spatula with a balance, to quickly and more accurately measure the samples. Particularly useful in ...
UOM:  1 * 1 items
Supplier:  OHAUS
Description:   A basic portable balance with an innovative design of draught shield.
Supplier:  OHAUS
Description:   Designed to be used in laboratory, industrial or educational applications - a balance that can be easily locked to be transported between sites of ope...
Supplier:  OHAUS
Description:   The CL series balance is a lightweight, portable scale perfectly suited for many applications including quality control, formulation, soil sampling, j...
Supplier:  OHAUS
Description:   Balance, Portable, Compact™, CS2000E, Readability: 1 g, Weighing cap.: 2000 g, Pan: 145×133 mm
Supplier:  OHAUS
Description:   The unique YA Series' weighing pan is integrated right into its carrying case, offering the ultimate combination of portability and protection. Just f...
Supplier:  OHAUS
Description:   Balance, Portable, Navigator, NVL511, Readability: 0,1 g, Weighing cap.: 510 g, Pan: 194×203 mm
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 8  of 18
  1  2  3  Next