Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "mechanical-balances"
https://ru-bot.vwr.com/stibo/smallweb/std.lang.all/40/54/18374054.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"9"
  List View Grid View Easy View (new)
Sort by:
Catalog Number: (611-5027)
Supplier:  OHAUS
Description:   Balance, Mechanical, Beam, 1119D, Readability: 1 g, Weighing cap.: 20 kg, Pan: Ø 279 mm
UOM:  1 * 1 items
Supplier:  OHAUS
Description:   Removable stainless steel pan, aluminium pressure cast base and beam.  These balances offer high precision and excellent value and are globa...
Supplier:  OHAUS
Description:   Durable triple beam mechanical balances. Ideal for precise weight determination and long-term use.
Supplier:  OHAUS
Description:   Balance, Mechanical, No tare single beam, 1450-SD, Readability: 0,1 g, Weighing cap.: 2000 g, Pan: Ø 152 mm
Catalog Number: (611-5357)
Supplier:  OHAUS
Description:   The Triple Beam Pro offers the convenience of a top loading balance while allowing your students to gain the understanding of measurement garnered thr...
UOM:  1 * 1 items
Supplier:  OHAUS
Description:   Balance, Mechanical, triple beam, 1650-00, Readability: 0,1 g, Weighing cap.: 610/2610 g, Pan: Ø 152 mm
UOM:  1 * 1 items
Catalog Number: (611-5365)
Supplier:  OHAUS
Description:   Simple 2-pan design with precision engineered beam for years of maintenance-free use in the classroom
UOM:  1 * 1 items
Supplier:  OHAUS
Description:   Balance, Mechanical, TJ2611, Readability: 0,1 g, Weighing cap.: 2610 g, Pan: 150 mm
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 8  of 9
  1  2  Next