Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • In Stock (2)
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "industrial-balances"
https://ru.vwr.com/stibo/smallweb/std.lang.all/40/52/18374052.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"47"
  List View Grid View Easy View (new)
Sort by:
Supplier:  VWR Collection
Description:   A robust, reliable series of balances for a wide range of applications in the laboratory, but also for general industrial use.
Supplier:  SARTORIUS BALANCES
Description:   The Cubis® range represents a modular concept from Sartorius to allow the customer to choose the components that suit the application area that is of ...
Supplier:  SARTORIUS BALANCES
Description:   Easy operation with monolithic weighing technology for users who want to perform precise weighing.
Supplier:  OHAUS
Description:   Large platform bench scales that incorporate touch screen, advanced weighing and proximity sensor technologies. The easy to use control unit can be mo...
Supplier:  OHAUS
Description:   A range of modular scales for tough industrial environments. Easy to use scales that are ideal for general weighing and simple counting applications i...
Supplier:  OHAUS
Description:   Balance, Industrial, Navigator, NVT10001, Readability: 0,5 g, Weighing cap.: 10 kg, Calibration: External weight adjustment
Supplier:  OHAUS
Description:   The Ranger 7000 series are rugged, high performance industrial scales for the toughest jobs. Ten advanced application modes can provide results for co...
Supplier:  OHAUS
Description:   Multipurpose, compact scale for producing precise results rapidly. Application modes include weighing, percent weighing, parts counting, accumulation ...
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 8  of 47
  1  2  3  4  5  6  Next