Determination of soil respiration, OxiTop® control B6/B6M
Supplier: WTW
OxiTop®
705-1010EA 5199 EUR
WTWA208232 WTWA208230 WTWA208231
Determination of soil respiration, OxiTop® control B6/B6M
Laboratory method for determining the microbial soil respiration according to DIN 19 737. Soil respiration measurements are used for forecasting, surveying and checking remediation work, for biodegradability measurements of substances (pesticides, fungicides, fertilisers etc.) as well as carrying out toxicity tests. This determination is possible with the OxiTop® Control System and special practically tested measuring vessels in a very accurate, simple and economical way.
  • Simple and precise
  • Cost effective
  • The measuring bottles and adapters can be autoclaved at 121 °C
  • Controller has a range of between 500 - 1350 hPa
  • The head flashes for five seconds when accessed by controller
Soil respiration measurements can be carried out in two different types of vessel. For actively respiring soils with strong CO₂ development, the MG1.0 measuring vessel is recommended with its large opening (~100 mm Ø) means that it can easily be used with large volume CO₂ absorber vessels for subsequent quantitative CO₂ determination.