Disinfectant wipes
115-8133EA 0 EUR
VWRI115-8133
Disinfectant wipes
Fleece wipes in dispenser, soaked in disinfecting solution. Optimum protection against fungi, bacteria and viruses.
  • For disinfection* of surfaces
  • For simple disinfection of hands
Kills bacteria: In accordance with EN 1040 and NF T 72-151. NF T 72-171. NF T 72-190
Kills fungi: In accordance with EN 1275
Kills viruses: In accordance with NF T 72-180 for polio, herpes and adenovirus, and in accordance with NF T 72-181 for T2, MS2
Colour: White
* Does not serve as a substitute for an immersion or sterilising bath