You Searched For: Wipes
415  results were found
SearchResultCount:"415"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (611-2886)
Supplier: VWR Collection
Description: Cleaning kit containing a puffer brush (lens cleaning brush), cotton cloth and pair of nitrile gloves.
UOM: 1 * 1 KIT
Supplier: CONTEC
Description: The VertiKlean Sponge is a disposable laboratory sponge. It consists of Polynit wiping material that has been laminated to a clean polyester foam. The...
Supplier: CONTEC
Description: Disposable Premira II Microfibre HighDuster covers reduce fibre shedding and provide effective dry and damp dusting for hard to reach areas. The micro...
Catalog Number: (BRDY018556)
Supplier: Brady
Description: This cleaning kit for printers includes: Instruction sheet, five cleaning cards soaked in IPA and the material safety data sheet.
UOM: 1 * 1 KIT
Catalog Number: (CONTMGMP0001)
Supplier: CONTEC
Description: These single use microfibre pads feature synthetic construction for the effective removal of both gross contamination and fine particles. Conveniently...
UOM: 1 * 140 items
Supplier: KIMBERLY CLARK
Description: Conventional high quality toilet rolls.
Supplier: Cytiva (Formerly Pall Lab)
Description: Handy dispenser kits allow one-handed dispensing of cellulose absorbent pads.
Catalog Number: (KIMB8441)
Supplier: KIMBERLY CLARK
Description: Provides the comfort of home.
UOM: 1 * 36 Roll
Catalog Number: (KIMB8408)
Supplier: KIMBERLY CLARK
Description: Premium toilet paper for any washroom.
UOM: 1 * 36 items
Catalog Number: (KIMB8042)
Supplier: KIMBERLY CLARK
Description: High quality toilet paper suitable for any washroom area.
UOM: 1 * 36 items
Catalog Number: (KIMB8511)
Supplier: KIMBERLY CLARK
Description: For reliability in busy washrooms that demand consistency, quality and value.
UOM: 1 * 6 Roll
Supplier: VWR Collection
Description: Suitable for cleaning various optical lenses such as objectives, binoculars, magnifying glasses, spectacles, as well as optical fibre filaments.
Supplier: VWR Collection
Description: These wipes are made from 100% continuous filament, double-ply, ultrasonically-sealed polyester to provide better absorbency. The wipe's size is perfe...
Supplier: VWR Collection
Description: These spunlace apertured wipes, made from 51% wood pulp and 49% polyester, are slightly abrasive on one side and soft on the other.
Sale
Supplier: VWR Collection
Description: These bleached, spunlace wipes, made from 55% wood pulp and 45% polyester, are extremely strong, even when wet.
Supplier: VWR Collection
Description: Absorbent, non woven wipe for use in ISO 5 and 6 cleanrooms.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 415
no targeter for Bottom