You Searched For: Weighing Papers
7  results were found
SearchResultCount:"7"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (516-0318)
Supplier: VWR Collection
Description: For weighing a range of substances (viscous products, powders, paste and liquids).
UOM: 1 * 250 items
Supplier: Whatman products (Cytiva)
Description: These Grade 2122 weigh paper sheets reliably allow samples to be transferred to scales without adding unwanted substances that could impact analytical...
Supplier: Whatman products (Cytiva)
Description: This weighing paper dissolves residue free in acid solution, which is critical in applications such as Kjeldahl analysis of nitrogen in soil.
Catalog Number: (HEATHS120116)
Supplier: HEATHROW
Description: These weighing papers are nitrogen-free for a very smooth surface and good sample recovery.
UOM: 1 * 500 items
Catalog Number: (611-9091)
Supplier: MACHEREY-NAGEL
Description: This is a transparent paper, smooth on both sides, which can be used as substitute for weighing boats. The smooth surface of the paper guarantees that...
UOM: 1 * 100 items
Catalog Number: (611-9420)
Supplier: NEOLAB
Description: Block pad with 250 precut sheets.
UOM: 1 * 250 items
Supplier: MAPSA
Description: Pergamyne paper, white, 40 g/m².
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 7 of 7
no targeter for Bottom