You Searched For: Watch Glasses
17  results were found
SearchResultCount:"17"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: Borosilicate 3.3 glass.
Supplier: VWR Collection
Description: PTFE.
Supplier: DWK Life Sciences
Description: DURAN®, borosilicate glass 3.3, clear, with fused edges.
Supplier: DWK Life Sciences
Description: Soda-lime glass.
Supplier: Bohlender
Description: Dish, made of PTFE, white.
Supplier: KARTELL
Description: PP, transparent.
Supplier: Brand
Description: PTFE, white.
Catalog Number: (PERCN9308003)
Supplier: PerkinElmer
Description: Disposable watch glasses to promote gentle refluxing. For use with 50 ml and 100 ml DigiTUBEs.
UOM: 1 * 1.000 items
Supplier: GLASWARENFABRIK KARL HECHT
Description: Evaporating dishes, made of glass, with fused rim.
Supplier: BOHEMIA CRISTAL HANDELSGES.MBH
Description: Watch glass dishes with fused edges made from soda-lime glass.
Supplier: BOHEMIA CRISTAL HANDELSGES.MBH
Description: Soda-lime glass.
Supplier: VITLAB
Description: Watch dishes, made of PTFE, white.
Supplier: KARTELL
Description: Made of PTFE, white.
Supplier: WITEG LABORTECHNIK GMBH
Description: Soda-lime glass.
Supplier: MURRAY, S & CO LTD
Description: Made of glass.
Supplier: SciLabware
Description: PYREX®, borosilicate glass, with fused edges.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 17
no targeter for Bottom