Cryoware labels, Nalgene®
Supplier: Thermo Fisher Scientific
Nalgene®
479-3291EA 70.5 EUR
479-3291
Cryoware labels, Nalgene®
Labels
Cloth labels for use at ultra-low temperatures, ballpoint pen compatible.
  • Will not shrink
For use at ultra-low temperatures, ballpoint pen compatible.
Dimensions: 25×50 mm.
Delivery information: 20 labels/sheet, 10 sheets/pack.