Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • In Stock (88)
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "cryogenic-vials"
https://ru.vwr.com/stibo/smallweb/std.lang.all/40/39/18374039.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"28"
  List View Grid View Easy View (new)
Sort by:
Supplier:  VWR Collection
Description:   Designed for the storage of biological material, human or animal cells, at temperatures as low as –196 °C (but should be used only in the gas phase of...
Supplier:  MATRIX TECHNOLOGIES CORP. LTD
Description:   Permanently attached, laser-etched 2D barcodes on the bottom of each tube allow you to scan and decode tubes at once without removing them from the ra...
Supplier:  BIOCISION, LLC
Description:   Each tube is printed with a unique barcode, graduation marks and a white writing surface. Tubes are autoclavable and produced from medical grade, chem...
Supplier:  Nalgene
Description:   Suitable for storage from general cold storage refrigeration (+4 °C) down to the vapour phase of liquid nitrogen.
Supplier:  MATRIX TECHNOLOGIES CORP. LTD
Description:   The tubes are ideal for sample collection and storage, including DNA, blood, cell lines and tissue samples. A permanent 2D barcode is encapsulated ont...
Supplier:  Nalgene
Description:   These storage tubes are ideally suited for research and general laboratory storage of high-value and genomic materials down to the vapour phase of liq...
Supplier:  NUNC
Description:   Vial, cryogenic, Nunc™, PPCO, Sterile, External thread, 0,5 ml, Ø ext.×H: 13×49 mm
Catalog Number: (MATRAB-1470)
Supplier:  MATRIX TECHNOLOGIES CORP. LTD
Description:   Unique and laser etched 2D barcode is encapsulated on the base to securely identify and track samples.
UOM:  1 * 240 items
Discontinued
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 8  of 28
  1  2  3  4  Next