Crimpers and decappers, Chromacol™
Supplier: Thermo Fisher Scientific
Chromacol™
548-1926EA 599 EUR
548-1926 548-1284 548-1292 548-1925 548-1923
Crimpers and decappers, Chromacol™
Crimpers and Decappers
Crimpers and decappers for 8 to 30 mm crimp top vials, with adjustable crimping height and pressure.
  • Chemically resistant surface finish
  • Easy and convenient handling
  • Robust jaws with hardened surface for long life