You Searched For: Chromatography Tubing
92  results were found
SearchResultCount:"92"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: Avantor
Description: Precision bore PEEK tubing is designed specifically for modern LC, LCMS and automation usage.
Supplier: Avantor
Description: Tubings are made of 316 stainless steel.
Supplier: Avantor
Description: PEEK
Supplier: Avantor
Description: PTFE
Catalog Number: (HICHHI-1097)
Supplier: Avantor
Description: The maximum operating temperature is 100 °C.
UOM: 1 * 1 items
Catalog Number: (HICHHI-1113)
Supplier: Avantor
Description: Tubing sleeve, PEEK, Avantor® Hichrom, Ø int.: 0,864 mm, Ø ext.: 1,575 mm, Colour: Blue
UOM: 1 * 1 items
Catalog Number: (HICHHI-1083)
Supplier: Avantor
Description: PEEKsil™ tubing is mechanically strong and has ideal characteristics for sealing with polymer fittings. It is comprised of high quality fused silica s...
UOM: 1 * 2 items
Supplier: Avantor
Description: Avantor® Hichrom, PEEK tubing, Black, Length: 1524 mm, Ø ext.: 1,5875 mm, Ø int.: 0,1016 mm
Supplier: Avantor
Description: 316 Stainless steel, precision bore ¹/₁₆" type tubing.
Catalog Number: (HICHHI-190)
Supplier: Avantor
Description: PEEK, Avantor® Hichrom, 90° Elbow
UOM: 1 * 1 items
Supplier: Avantor
Description: Tubings are made of 316 stainless steel.
Supplier: Avantor
Description: 316 Stainless steel, precision bore ¹/₁₆" type tubing.
Catalog Number: (HICHHI-171)
Supplier: Avantor
Description: PEEK.
UOM: 1 * 10 items
Catalog Number: (HI-259)
Supplier: Avantor
Description: Avantor® Hichrom, Tubing, stainless steel, Length: 0,15 m, Ø ext.: 3,18 mm
UOM: 1 * 1 items
Supplier: SGE a Trajan company
Description: Precision bore PEEK tubing is designed specifically for modern LC, LCMS and automation usage.
Supplier: SGE a Trajan company
Description: GLT™ (Glass Lined Tubing) is made by fusing a borosilicate glass lining onto the inside surface of stainless steel tubing. Ideal for HPLC columns, esp...
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 92
no targeter for Bottom