Drying tubes with Quickfit® ground joints, U-shaped
Supplier: SciLabware
BIBBMF21/2EA 227.69 EUR
BIBBMF21/2
Drying tubes with Quickfit® ground joints, U-shaped
PYREX®, borosilicate glass. Two ground sockets.
  • Excellent chemical resistance
  • High temperature resistance
For use with desiccants, such as calcium chloride.