PCR tubes, 0,5 ml, with flat or domed cap, ABgene®
Supplier: Thermo Fisher Scientific
ABgene®
732-0061EA 92 EUR
732-0061 732-4804
PCR tubes, 0,5 ml, with flat or domed cap, ABgene®
Tubes PCR Tubes
PP.
Suitable for 0,5 ml thermal cycler blocks.
  • 0,6 ml maximum tube capacity when closed
  • Integral snap-shut cap
  • Certified free from DNase, RNase and Human DNA
Suitable for 0,5 ml thermal cycler blocks.
  • 0,6 ml maximum tube capacity when closed
  • Integral snap-shut cap
  • Certified free from DNase, RNase and Human DNA
Colour: Clear