Microcentrifuge tubes, 2 ml, SuperClear
VWR® SuperClear™
211-0118EA 30.8 EUR
VWRI211-0118 VWRI211-0038 VWRI211-0119 VWRI211-0034
Microcentrifuge tubes, 2 ml, SuperClear
Ultra-clear PP, graduated, autoclavable and freezable with flat cap. Amber coloured for light-sensitive substances.
  • Can withstand centrifugation to 20000 ×g
  • Fit all standard, high capacity rotors
  • Tubes have frosted side labelling surface and flat cap can be easily labelled and pierced
  • Every lot is tested and guaranteed DNase- and RNase-free
  • Sustainable design based on virgin resins
Packaging: In resealable bag. Assorted colours pack includes red, blue, green, yellow and orange tubes in separate 100 tube bags.