Microcentrifuge tubes, 0,65 ml, SuperClear boil-proof
211-0031EA 0 EUR
VWRI211-0031
Microcentrifuge tubes, 0,65 ml, SuperClear boil-proof
Ultra-clear PP, graduated, autoclavable and freezable with flat caps.
  • Withstand centrifugation to 30000×g
  • Fit all standard, high capacity rotors
  • Flat cap can be easily labelled and pierced, includes a special 90° hinge, making it easier to close while pipetting one-handed
  • Every lot is tested and guaranteed DNase- and RNase-free
  • Sustainable design based on virgin resins
Packaging: In resealable bags. Assorted colours pack includes red, blue, green, yellow and orange tubes in separate 200 tube bags.