You Searched For: Centrifuge Tubes
136  results were found
SearchResultCount:"136"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: PTFE, round bottom, smooth surface, without screw cap.
Supplier: VWR Collection
Description: PTFE, smooth surface, with or without screw cap.
Supplier: VWR Collection
Description: These leakproof, conical bottom centrifuge tubes prevent breakage and leakage during high speed centrifugation, and are made of non cytotoxic, medical...
Sale
Supplier: VWR Collection
Description: These leakproof disposable conical bottom and free-standing tubes feature black graduations and smooth inner walls for easy filling and sample prepara...
Sale
Supplier: VWR Collection
Description: Reusable centrifuge tubes made from borosilicate 3.3 glass, available with round or conical bottom, graduated or non graduated.
Supplier: VWR Collection
Description: Centrifuge tubes are ideal for use in centrifuging samples or storage in many research areas.
Supplier: VWR Collection
Description: Available with round or conical bottom, graduated or non graduated. Not suitable for applications related with strong heat or rapid temperature change...
Sale
Supplier: VWR Collection
Description: Made from borosilicate 3.3 glass, with heavy wall to withstand higher speeds in centrifugation.
Supplier: VWR Collection
Description: Manufactured of high-quality USP Class 6 gamma resistent clear polypropylene with HDPE screw cap.
Catalog Number: (525-0155)
Supplier: VWR Collection
Description: These popular, conical-bottom centrifuge tubes are designed for high speed centrifugation, and are made of non cytotoxic, medical grade resin that all...
UOM: 1 * 500 items
Low Manufacturing Impact
Sustainable Packaging
Supplier: VWR Collection
Description: These leakproof centrifuge tubes are manufactured from dark amber PP, providing protection for light-sensitive samples by blocking UV light.
Supplier: VWR Collection
Description: PP, high grade gamma resistance, conical, with HDPE flat cap.
Sale
Supplier: VWR Collection
Description: PS, clear, conical, with flat cap with integral sealing.
Supplier: VWR Collection
Description: Made of high quality, USP class 6 gamma-resistent clear polypropylene with HDPE screw cap.
Supplier: VWR Collection
Description: These 5 ml microtubes are made of transparent PP with a snap flat cap, easy to be opened and closed.
Supplier: VWR Collection
Description: These leakproof centrifuge tubes are ideal for environmental uses, such as water analysis and other applications wherein tubes have the potential to c...
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 136
no targeter for Bottom