Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • In Stock (7)
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "Capillary Tubes"
https://ru-bot.vwr.com/stibo/smallweb/std.lang.all/93/27/18989327.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"19"
  List View Grid View Easy View (new)
Sort by:
Catalog Number: (MODU120013)
Supplier:  VITREX MEDICAL A/S
Description:   Sedicaps capillary tubes with ringmarks at 100/102 mm.
UOM:  1 * 1.000 items
Supplier:  WILMAD/LABGLASS/LUREX MFG
Description:   Capillary Tubes, for NMR measurement, Pyrex® glass, Ø ext.: 1,0 mm, Ø int.: 0,8 mm, Length: 203 mm
Supplier:  HIRSCHMANN
Description:   Ringcap, for reflotron, disposable capillary pipette with a ringmark at 32 µl, Li-heparinized, DURAN, conformitycertified. 1 * 500 items
UOM:  1 * 500 items
Supplier:  HIRSCHMANN
Description:   Capillary tube, both ends open, Borosilicate glass, Ø ext.: 1,35 mm, Ø int.: 0,95 mm, Length: 100 mm
Supplier:  HIRSCHMANN
Description:   Capillaries, Na-heparinized (coated) , for blood gas analysis, Borosilicate glass 3.3, Ø ext.: 1,75 mm, Ø int.: 1,35 mm, Length: 75 mm, Nominal volume: 100 µl
Supplier:  MARIENFELD
Description:   Capillaries, Na-heparinized (coated) , for blood gas analysis, Glass, Ø ext.: 1,75 mm, Length: 75 mm, Nominal volume: 82,5 µl
Supplier:  MARIENFELD
Description:   Tube capillary 60?l 0.9-1mm sodium-heparinized microhaematocrit 1 * 1.000 items
Catalog Number: (MARI3231000)
Supplier:  MARIENFELD
Description:   Hematocrit tubes, acc. to Wintrobe, with double numbered scale in red / white colour, Soda-Lime glass, Ø ext.: 6,5 mm, Ø int.: 2,9 mm, Length: 105 mm
UOM:  1 * 20 items
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 8  of 19
  1  2  3  Next