You Searched For: Tripods
6  results were found
SearchResultCount:"6"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: USBECK Laborgeräte
Description: Tripod, Stainless steel, Int.Ø×H: 120×250 mm
Supplier: BOCHEM
Description: Tripods made from stainless steel or tempered cast iron (zincked).
Supplier: RSG
Description: Separable tripods made from stainless steel.
Catalog Number: (769-1150)
Supplier: Industrial Physics
Description: The spirit level is easy to balance thanks to the adjustable feet of the unit.
UOM: 1 * 1 items
Catalog Number: (HECH41091010)
Supplier: GLASWARENFABRIK KARL HECHT
Description: These tripods are made of galvanised iron.
UOM: 1 * 1 items
Supplier: JUCHHEIM Laborgerate GmbH
Description: Stainless steel.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 6 of 6
no targeter for Bottom