Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
Cutters
https://ru.vwr.com/stibo/smallweb/std.lang.all/50/93/18945093.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"32"
  List View Grid View Easy View (new)
Sort by:
Supplier:  Hichrom Limited
Description:   The Clean-cut™ cutter is designed specifically for cutting polymeric HPLC tubing from 1/16” to 1/8” with no distortion of cross-section and without cl...
Catalog Number: (WHEA177105)
Supplier:  DWK Life Sciences
Description:   These snapers for ampoules protect against glass splinters and prevent cross contamination.
UOM:  1 * 144 items
Catalog Number: (18-1112-46)
Supplier:  GE Healthcare - Life Sciences
Description:   Tubing cutter
UOM:  1 * 1 items
Catalog Number: (590-0850)
Supplier:  SCAT Europe
Description:   This cutter is made of PP, blue, with stainless steel blade.
UOM:  1 * 1 items
Supplier:  SGE a Trajan company
Description:   Cutter for steel tubing.
Supplier:  ENTEGRIS INC
Description:   CUTTER - PIPE AND TUBE 1/8 - 1 5/8 OD 213-30-1 1 * 1 items
Supplier:  BRADY & CO LTD, W. H.
Description:   These hole punches with the design of tongs, 50 mm wide which allows to punch the middle of the cardsup to 100 mm wide.
Catalog Number: (BOHLS1852-28)
Supplier:  Bohlender
Description:   Ideal for cutting plastic and rubber tubing with and without textile reinforcement.
UOM:  1 * 1 items
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 8  of 32
  1  2  3  4  Next