Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
Tools
SearchPresentationType-VERTICAL
SearchResultCount:"5"
  List View Easy View (new)
Sort by:
Catalog Number: (WHEA177105)
Supplier:  DWK Life Sciences
Description:   These snapers for ampoules protect against glass splinters and prevent cross contamination.
UOM:  1 * 144 items
Supplier:  CAMAG
Description:   Ideal instrument for exact cutting of TLC/HPTLC plates that are frequently used in method development and pre-testing. The smartCut can cut glass plat...
Catalog Number: (BELAF389520000)
Supplier:  Bel-Art
Description:   The box style design of the ampoule opener allows one handed opening of up to 3 ampoules at a time by simply pressing down on lid. Ampoule necks are s...
UOM:  1 * 1 items
Supplier:  SciLabware
Description:   Ampoule breakers, HDPE, for 5 - 10 ml glass ampoules
UOM:  1 * 1.000 items
Supplier:  Hach
Description:   These breakers for ampoules are designed to prevent contamination. Breaker snaps ampoule tip off.
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 5  of 5
A new look and a renewed commitment
Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.
Learn More Start Shopping