VWR®, Temperature Indicating Strips
Supplier: VWR
VWR®
620-9103EA 33.6 EUR
620-9103 620-9104 620-9105 620-9106 620-9102
VWR®, Temperature Indicating Strips
Temperature Indicating Strips
These temperature indicating strips are designed for rapid measurements where conventional thermometers are impractical. They are suitable for use with equipment such as heating chambers and products which are sensitive at high temperatures.
  • Fast response within 1 second
  • Self-adhesive and irreversible - six points
  • Storage at maximum 25 °C for up to 2 years