Pocket thermometers
Supplier: Amarell
620-6738EA 0 EUR
AMARG13000/G13800 AMARG13214/G13800
Pocket thermometers
These standard maximum mercury thermometers display and retain recorded high temperatures until reset. Resetting can be done with a magnet.
  • Total or partial immersion
  • Enclosed scale and solid stem
  • Colourless capillary
Until 10th October 2017 it is still allowed to use mercury thermometers if they are exclusively intended to be used for tests as per standards which require the usage of mercury thermometers.
Delivery information: Supplied with magnets.
Caution: At a use of more than 200 °C, please take into account the instructions for use (supplied with thermometer).