Pocket thermometers
Supplier: Amarell
620-1747EA 21.22 EUR
AMARG15404
Pocket thermometers
Extra comfort and security. The thermometers with red liquid filling have a colourless capillary.
  • Pen size
  • Total immersion
  • Enclosed scale
Delivery information: All pocket thermometers supplied in nickel plated double receptacle.