Contact thermometers
Supplier: Ludwig Schneider
620-4152EA 0 EUR
620-4152 620-4153 620-4154 620-4132 620-4146
Contact thermometers
Thermometers Analogue Thermometers
These thermometers are designed for tough applications. They have two scales; in the upper part is the setting scale and in the lower part the indication or temperature scale. The setting of the temperature by means of a special contact wire which can be moved up or down the temperature scale.
  • The temperature setting is adjusted by rotating part of the cap which in turn moves a threaded spindle
  • Rapid response to temperature changes in approximately 1 to 1,2 seconds in standard vessels
  • Threaded spindle in stainless steel for smooth running of the setting mechanism
  • Switching operation inside a hermetically closed space filled with inert gas
Filling: Mercury
D.B.P.