High precision thermometers
Supplier: VWR Collection
620-0662EA 0 EUR
VWRF09654.407 VWRF09639.212 VWRF09640.273 VWRF09637.102 VWRF02913.100 VWRF09638.082 VWRF09638.162 VWRF09638.025 VWRF09638.127 VWRF09630.030 VWRF09656.164 VWRF09637.044 VWRF09637.088 VWRF09638.207 VWRF09637.066 VWRF09637.011 VWRF09637.022 VWRF09656.255 VWRF09654.065 VWRF09654.167
High precision thermometers
These mercury thermometers are designed for highly accurate measurements and calibration of other thermometers. The graduations allow for an easier reading.
  • Total immersion
Until 10th October 2017 it is still allowed to use mercury thermometers if they are exclusively intended to be used for tests as per standards which require the usage of mercury thermometers.
Certifications: Conform to NF B 35502.