Certified thermometers
Supplier: Amarell
620-0992EA 91.09 EUR
AMARA301770 AMARA300560 AMARA300440 AMARA301650 AMARA300320 AMARA301410 AMARA301014 AMARA300960 AMARA300840 AMARA300720 AMARA300600 AMARA301380 AMARA300170 AMARA300690 AMARA300570 AMARA301000 AMARA300430 AMARA301400 AMARA301368 AMARA300520KB AMARA300798 AMARA301490 AMARA300280 AMARA300160 AMARA301250 AMARA301130 AMARA300540 AMARA301630 AMARA300420 AMARA300300 AMARA301510 AMARA300664 AMARA300940 AMARA301358 AMARA300820 AMARA300788 AMARA300210CC3 AMARA300150 AMARA300030 AMARA300010 AMARA300770 AMARA301500 AMARA300654 AMARA301348 AMARA300260F AMARA301190 AMARA301590 AMARA300380 AMARA303771 AMARA301230 AMARA301330 AMARA301210 AMARA300880 AMARA300520 AMARA301730 AMARA300400 AMARA301610 AMARA300644 AMARA300920 AMARA300808 AMARA301340 AMARA300130 AMARA301440 AMARA300210F AMARA300230 AMARA301320 AMARA300110 AMARA300750 AMARA301048 AMARA300634 AMARA301420CC4 AMARA301290 AMARA301170 AMARA300030DKD4 AMARA301690 AMARA300360 AMARA301570 AMARA301450 AMARA300240 AMARA300340 AMARA301550 AMARA301430 AMARA303211 AMARA301034 AMARA300980 AMARA301310 AMARA300860 AMARA300620 AMARA301830 AMARA300500 AMARA301710 AMARA300900 AMARA300210DKD13 AMARA300190 AMARA300070 AMARA301160 AMARA300590 AMARA301560 AMARA300450 AMARA301420 AMARA301300 AMARA301940 AMARA300730 AMARA300610 AMARA300150DKD4 AMARA301390 AMARA301270 AMARA301150 AMARA301790 AMARA300580 AMARA300460 AMARA301670
Certified thermometers
These models are suitable for government verification or calibration.
These models are suitable for government verification or calibration.
  • White backed
Filling: mercury, toluene or mercury-thallium alloy
  • Most are mercury filled
  • White backed
  • Compliant with ASTM
Until 10th October 2017 it is still allowed to use mercury thermometers if they are exclusively intended to be used for tests as per standards which require the usage of mercury thermometers.
Until 10th October 2017 it is still allowed to use mercury thermometers if they are exclusively intended to be used for tests as per standards which require the usage of mercury thermometers.
Certifications: Compliant with ASTM.