You Searched For: Swabs
157  results were found
SearchResultCount:"157"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: Critical swabs designed for cleaning disc drives, optics, PC board assemblies and other sensitive components.
Sale
Supplier: VWR Collection
Description: Collection and transport swabs for bacterial specimens with applicators and transport media.
Supplier: VWR Collection
Description: Disposable 'foam over cotton' tip swab features 100% polyurethane foam with 100 ppi, and a wooden shaft.
Supplier: VWR Collection
Description: Swabs feature 100 ppi open or closed cell polyurethane foam, with 100% virgin polypropylene handle. Some have a blue, glass fibre, reinforced polyprop...
Supplier: VWR Chemicals
Description: Surface sampling sponges in two forms: Dry sponge sterile sampling bag with write-on strip, containing a dry, biocide-free cellulose sponge, or wet sp...
MSDS Sale
Catalog Number: (710-0442)
Supplier: VWR Collection
Description: Transport swab with plastic shaft, synthetic tip, and Amies agar gel.
UOM: 1 * 50 items
Catalog Number: (COPA414C)
Supplier: COPAN
Description: The M40 Transystem line is the latest generation of traditional bacteriology transport swabs. It was designed to provide the ultimate performance capa...
UOM: 1 * 50 items
Supplier: Texwipe
Description: These Foamtipsâ„¢ swabs are constructed from medical grade polyurethane foam, which features the best open cell structure, providing particle entrapment...
Supplier: COPAN
Description: A range of collection and transport devices for bacterial specimens.
Supplier: COPAN
Description: Liquid transport medium is contained in a sponge carrier located in the bottom of the tube.
Supplier: THERMO FISHER DIAGNOSTICS GMBH
Description: Effective transportation of throat, vaginal, wound and skin swabs conforming to highest medical device classification - Class II a surgically invasive...
Catalog Number: (MMMPRS96010BPW)
Supplier: 3M Food Safety
Description: 3Mâ„¢ Swab-Sampler with 10 ml Buffered Peptone Water Broth is a convenient and quick collection device for environmental sampling and testing over small...
UOM: 1 * 100 items
Supplier: COPAN
Description: Surface control swabs and sponges designed for the hygiene monitoring of surfaces.
Supplier: COPAN
Description: Swab rinse kits (SRK®) for microbiological examination of surfaces.
Supplier: Texwipe
Description: Made from polyurethane foam with 100 pore per inch porosity.
Supplier: Texwipe
Description: Swab features a compressed PU head with precision pointed tip and side ridges, on a short PP handle.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 157
no targeter for Bottom