VWR® Thermoplate, Surface Protection Plates
Supplier: VWR
115-0097EA 149 EUR
VWRI115-0097
VWR® Thermoplate, Surface Protection Plates
These plates made from ceramic glass protect high quality surfaces from high temperature containers such as crucibles, evaporating dishes, glass containers.
  • Temperature resistant from −235,5 to +648,8 °C
  • Easily cleaned, dirt-repellent surface
  • Low thermal expansion, resistant to thermal shocks
  • Reliable protection against deep frozen items
  • Feet at the edges of the plates prevent direct contact with the surfaces to be protected