You Searched For: Surface Protectors
47  results were found
SearchResultCount:"47"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: High quality, smooth, absorbent paper that quickly absorbs liquid spills with a thick, laminated polyethylene layer that prevents seeping onto the wor...
Catalog Number: (444-0516)
Supplier: VWR Collection
Description: Bench protector, VWR®, Spare dimpled mat
UOM: 1 * 1 items
Catalog Number: (631-1059)
Supplier: VWR Collection
Description: Highly absorbent paper sheets for bench and surface protection.
UOM: 1 * 25 items
Supplier: VWR Collection
Description: Bench protector, VWR®, Rubber mat, 330×279 mm
Catalog Number: (111-2431)
Supplier: VWR Collection
Description: These durable FDA food grade silicone lab mats are ideal for every type of laboratory and more specialised areas such as cold rooms, cleanrooms, steri...
UOM: 1 * 1 items
Supplier: DESCO
Description: ESD workstation cover protects the workstation from ESD, dust and other contaminants.
Sale
Supplier: Bohlender
Description: For use as a table pad, slideway or for insulation.
Supplier: Bohlender
Description: Protection sheets made from PTFE or FEP.
Catalog Number: (KIMB7093)
Supplier: KIMBERLY CLARK
Description: Protective sheets, water repellant, Kimcare®, 375×500 mm
UOM: 1 * 100 items
Supplier: Thermo Fisher Scientific
Description: Made from 3,2 mm thick closed-cell polyethylene foam this bench liner provides cushioning and liquid barrier protection.
Catalog Number: (KIMB7398)
Supplier: KIMBERLY CLARK
Description: This two ply couch cover is strong, tear resistant and durable.
UOM: 1 * 12 Roll
Supplier: Thermo Fisher Scientific
Description: Versi-Dry lab soakers protect surfaces and equipment, cushion breakable items and quickly absorb spills. The polythene backing is waterproof and resis...
Catalog Number: (KIMB7396)
Supplier: KIMBERLY CLARK
Description: SCOTT® couch covers are strong, tear resistant and durable.
UOM: 1 * 12 Roll
Supplier: Sartorius
Description: The LabSorb protection papers are highly absorptive papers coated on one side with polyethylene.
Supplier: Whatman products (Cytiva)
Description: Cytiva's Whatman™ Benchkote® Plus bench protector is a thicker, more absorbent version of the original Benchkote® bench paper. Designed for demanding ...
Supplier: DEUTSCH & NEUMANN
Description: Black protection sheets made from fluoroelastomer rubber (FPM). Durometer 75 Shore A.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 47
no targeter for Bottom