VWR®, Cork Rings for Round Bottom Flasks
Supplier: VWR
VWR®
217-1003EA 54.8 EUR
217-1003 217-1002 217-1001 217-1000 217-0810 217-1005 217-1004
VWR®, Cork Rings for Round Bottom Flasks
Supports Flask Supports
The ideal rest for round bottom flasks and other round bottom glass vessels. Compressed cork, natural.
  • Economical and from a renewable resource
  • Lightweight, tough
  • Good thermal resistance