You Searched For: Supports
28  results were found
SearchResultCount:"28"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: Made of natural cork.
Supplier: VWR Collection
Description: The ideal rest for round bottom flasks and other round bottom glass vessels. Compressed cork, natural.
Supplier: Saint-Gobain
Description: Can be used to provide a stable support for round-bottomed flasks and other containers with curved bases. Reversible. Non-slip, soft, stackable and ea...
Supplier: ROBU GLASFILTER
Description: Conical gaskets, made of rubber, to place between filter funnel and filter flask.
Supplier: ROBU GLASFILTER
Description: Rubber sleeves for filter crucibles.
Supplier: KGW (KARLSRUHER GLASTECHN.)
Description: Tabletop supporting frame, pipe Ø 27 mm, 800 mm height, for: flat flanges DN 150
Supplier: DEUTSCH & NEUMANN
Description: Rubber, autoclavable.
Supplier: DEUTSCH & NEUMANN
Description: Natural rubber.
Supplier: DEUTSCH & NEUMANN
Description: These sleeves for filter crucibles are made from red natural rubber.
Supplier: USBECK Laborgeräte
Description: Flask holder, Cork, For: 500 - 1000 ml, Ø ext.: 140 mm, Int.Ø bottom: 90 mm, Height: 30 mm
Catalog Number: (IKAA4448400)
Supplier: IKA
Description: This flask carrier with handle is constructed from anodised aluminium.
UOM: 1 * 1 items
Supplier: Corning
Description: PP tubes, polyetherimide support cushions.
Catalog Number: (AZLOTWR038)
Supplier: SciLabware
Description: This stand is suitable for most round bottomed flasks.
UOM: 1 * 1 items
Catalog Number: (KART420)
Supplier: KARTELL
Description: Set of 7 adapters, conical, made of black neoprene, sleeve supports for Büchner funnels and filter flasks.
UOM: 1 * 1 items
Supplier: GLASWARENFABRIK KARL HECHT
Description: Flask holder, Cork, For: 10 - 100 ml
Catalog Number: (214-1920)
Supplier: KARTELL
Description: Provide secure, non-slip support for round bottom flasks and any other flasks with convex bases of any height. The concentric inner rings can hold fla...
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 28
no targeter for Bottom