Rubber stoppers
Supplier: Saint-Gobain
217-0729EA 121 EUR
VERN203030 VERN203050 VERN203023 VERN203042 VERN203046 VERN203039
Rubber stoppers
Conical natural grey-blue rubber stoppers with one hole.
  • Resistant to all common laboratory chemicals
  • Provides a perfect seal and exceptional bonding
  • Resistant to temperatures from −5 to +70 °C (120 °C in humid atmosphere)
  • Light sensitive product, do not store in direct sunlight
Conical stopper drilled, highly elastic, for perfect sealing and adhesion. One hole with different diameter according the pattern. Soft and robust for a complete leakproof & grip. Store and prevent from light.
Certifications: DIN 12871