You Searched For: Stoppers
124  results were found
SearchResultCount:"124"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: These plugs are made of bonded dense weave cellulose acetate which provides an excellent barrier to mite infestation. They are precision manufactured ...
Supplier: VWR Collection
Description: Stopper for test tubes Ø 12 to 16 mm, polyethylene, with tongues.
Supplier: VWR Collection
Description: These stoppers are made of cellulose and are used for microbiological samples and tissue cultures in test tubes or Erlenmeyer flasks.
Supplier: VWR Collection
Description: Greaseless closure, which prevents ground joints from seizing. With inclined, circumferential sealing lips for better sealing and reduced risk of brea...
Supplier: VWR Collection
Description: Borosilicate 3.3 glass.
Sale
Supplier: VWR Collection
Description: Natural cork, A1 quality.
Sale
Catalog Number: (217-0895)
Supplier: VWR Collection
Description: Stopper, cork
UOM: 1 * 20 items
Catalog Number: (462-0585)
Supplier: VWR Collection
Description: Drain stoppers, VWR®
UOM: 1 * 5 items
Supplier: Simport Scientific
Description: HDPE. Fit tubes of 15 ml capacity and 17 mm Ø.
Supplier: DWK Life Sciences
Description: This bromobutyl rubber closure provides a gas-tight seal for all GL 25 and GL 45 laboratory bottles with sizes from 10 to 20000 ml.
Supplier: Lenz Laborglas GmbH & CO.KG
Description: These standard taper hollow stoppers are made from borosilicate glass 3.3 and have a hexagonal head.
Supplier: Bohlender
Description: For closing ground joint parts. With ground joint and sealing rings on the outside; knurled or hexagonal grip. Compared with glass stoppers, they are ...
Supplier: Freudenberg Medical Europe GmbH
Description: HelixMark silicone stoppers enable reliable sealing of glass tubes, vials and flasks. The stoppers are suitable for all commonly used diameters and si...
Sale
Supplier: Simport Scientific
Description: PE Plug type push-in caps and 2-position snap caps are made of polyethylene for test tubes with outside diameters of 12 or 17 mm. Dual position caps o...
Supplier: Lenz Laborglas GmbH & CO.KG
Description: DURAN®, borosilicate glass stopper, clear, solid or semi hollow.
Supplier: Saint-Gobain
Description: These stoppers are made of translucent silicone. The stoppers guarantee absolute impermeability, even when the edges of the tubes or glass, metal or p...
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 124
no targeter for Bottom