You Searched For: Stopcocks
130  results were found
SearchResultCount:"130"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (467-0111)
Supplier: VWR Collection
Description: Spare stopcock, VWR®, Stopcock with PTFE spindle, NS 24/29
UOM: 1 * 1 items
Supplier: Avantor Fluid Handling
Description: Exceptional durability.
Catalog Number: (216-1745)
Supplier: VWR Collection
Description: PP, white/purple.
UOM: 1 * 1 items
Catalog Number: (612-7206)
Supplier: VWR Collection
Description: VWR®, Spare stopcock for burettes 612-6694, 612-6695 and 612-6696
UOM: 1 * 1 items
New Product
Supplier: Lenz Laborglas GmbH & CO.KG
Description: Vacuum stopcocks, DURAN® with side arms and hollow NS glass plug.
Supplier: Lenz Laborglas GmbH & CO.KG
Description: DURAN®, borosilicate glass, with solid glass or PTFE key.
Supplier: Bohlender
Description: PTFE.
Supplier: Bohlender
Description: Made of PTFE.
Supplier: DWK Life Sciences
Description: Borosilicate glass 3.3, clear, with PTFE spindle, for GL 32 screw thread.
Supplier: Bohlender
Description: Miniature stopcocks of PTFE with male or female connection threads and stopcock plug of FEP.
Supplier: ENTEGRIS INC
Description: Two- or three way, ¹/₈" orifice.
Supplier: Bohlender
Description: PTFE, white.
Supplier: Bohlender
Description: Made of PTFE/PPS.
Supplier: Bohlender
Description: Made of PTFE.
Supplier: Lenz Laborglas GmbH & CO.KG
Description: Plugs, Borosilicate glass, Hole Ø: 1,5 mm, Spigot: 12,5 NS, For: Stopcocks, straight bore, Spare solid glass stopcock key
Catalog Number: (BOHLJ1265-10)
Supplier: Bohlender
Description: PTFE/PP.
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 130
no targeter for Bottom