VWR®, Magnetic Stirring Bar Retrievers
Supplier: VWR
VWR®
VWRU89026-260EA 0 EUR
VWRU89026-260 442-0526 442-0523 442-0524 442-0525 VWRU89026-266 VWRU89026-256 VWRU89026-264 VWRU89026-254 VWRU89026-262
VWR®, Magnetic Stirring Bar Retrievers
Stirrers Stirring Tools
PTFE covered, strong Alnico V magnetic core. For the removal of stirring magnets from vessels of all kinds.
  • Excellent chemical resistance
  • Temperature resistant, suitable for use at high and low temperatures (−200 to +280 °C)