VWR®, Magnetic Stirrers, with Crosshead
Supplier: VWR
VWR®
442-0416EA 66.5 EUR
442-0416 442-0417 442-0418 442-0419 442-0412 442-0413 442-0414 442-0415 442-0420 442-0421 442-0411 442-0422
VWR®, Magnetic Stirrers, with Crosshead
Stirrers Magnetic Stirring Bars
PTFE covered, strong Alnico V magnetic core. Smooth surface. Particularly suitable for small stirring vessels, extremely effective stirring action.
  • Excellent chemical resistance
  • Temperature resistant, suitable for high and low temperature applications (−200 to +280 °C)