VWR®, Magnetic stirring bars, oval
Supplier: VWR
VWR®
442-0405EA 99.1 EUR
442-0405 442-0406 442-0407 442-0408 442-0401 442-0402 442-0403 442-0404 442-0410 442-0409
VWR®, Magnetic stirring bars, oval
Stirrers Magnetic Stirring Bars
PTFE covered, Alnico V magnetic core. Smooth surface. Designed for use in round bottomed flasks.
  • Excellent chemical resistance
  • Temperature resistant, suitable for high and low temperature applications (−200 to +280 °C)