Safety spectacles, Teknic
Supplier: Bollé Safety
111-0339EA 20.6 EUR
BOLETEKCI
Safety spectacles, Teknic
Spectacles with inbuilt upper, lower and side protection guards.
  • Extra large integrated side shield
  • Non slip nosepiece and adjustable temple length
  • Screw frame for easy lens replacement
  • Optional braided neck cord
Weight: 45 g
Certifications: EN 166, EN 170