You Searched For: Spatulas
220  results were found
SearchResultCount:"220"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: PP. Multi-purpose, robust, inexpensive tools for collecting, transferring and processing samples of chemical and biological materials.
Catalog Number: (231-0492)
Supplier: VWR Collection
Description: PS, white. This spatula, with its long, ergonomic solid handle and pointed blade, can easily pierce paper and plastic bags.
UOM: 1 * 100 items
Supplier: RSG
Description: Stainless steel 18/10, anti-magnetic.
Supplier: RSG
Description: Rigid micro double spatulas made of stainless steel.
Supplier: RSG
Description: Chromium steel, hardened, extremely flexible blade.
Supplier: RSG
Description: Pharmacist's spatulas with stainless steel blade and black plastic handle.
Catalog Number: (RSGA316.150)
Supplier: RSG
Description: Stainless steel 18/10. Micro spatula with one square end.
UOM: 1 * 10 items
Supplier: RSG
Description: Spatula, Stainless steel 18/10, Non sterile, Length: 150 mm
Supplier: RSG
Description: Stainless steel 18/10, anti-magnetic. Multipurpose spoon spatula for a wide range of uses.
Supplier: RSG
Description: Stainless steel 18/10, anti-magnetic.
Supplier: RSG
Description: Stainless steel, 18/10. Micro spatulas with black plastic handle.
Supplier: BÃœRKLE
Description: The sampling palette knife spatula can be used to apply, distribute or evenly spread viscous media such as pastes and creams. The palette knife spatul...
Supplier: Bohlender
Description: PTFE, white.
Supplier: BÃœRKLE
Description: PS. Double spatula with a 0,5 ml spoon and a 17 mm spatula blade. Ideal for use as a sampling spoon, for measuring or transferring small amounts, as a...
Supplier: RSG
Description: These are made of stainless steel 18/10.
Supplier: BÃœRKLE
Description: The sampling palette knife spatula can be used to apply, distribute or evenly spread viscous media such as pastes and creams. The palette knife spatul...
Low Manufacturing Impact
Product Transparency and Disclosure
Sustainable Materials
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 220
no targeter for Bottom