You Searched For: General Purpose Sealing Films
13  results were found
SearchResultCount:"13"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: HEATHROW
Description: Parafilm® M is a semi-transparent, flexible, thermoplastic film with two-way stretch. This odourless and colourless film provides a tight, moisture-pr...
Catalog Number: (BRDY065508)
Supplier: Brady
Description: Sealing film, Polyester
UOM: 1 * 40 items
Catalog Number: (113-9674)
Supplier: HEATHROW
Description: Universal, semi-transparent, highly elastic film designed for airtight and moisture-proof sealing and many other manual applications. The film can be ...
UOM: 1 * 1 items
Supplier: LONZA
Description: GelBond® film is a transparent, flexible polyester film designed to support agarose gels. Gels cast on GelBond® film remain permanently attached to th...
Supplier: LONZA
Description: GelBond® PAG film is a transparent, flexible polyester film designed to support polyacrylamide or MDE™ gels. Gels remain permanently attached to the f...
Supplier: Brand
Description: Seal-R-filmâ„¢ sealing film seals beakers, flasks, and other vessels.
Supplier: DIVERSIFIED BIOTECH
Description: Sealing film, DuraSeal®, Length: 152,4 m, Width: 50,8 mm, PE
Supplier: LOSCH, RAINER
Description: Sealing film, Length: 100 m, Width: 300 mm, PE
Sale
Catalog Number: (FISH2N600600)
Supplier: FISHER CONTAINER CORP
Description: Clear LDPE, amine-free antistatic film ideal for partitioning during retrofitting or construction. Specially formulated to reduce particulate contamin...
UOM: 1 * 1 items
Catalog Number: (DIVEBMTM-2000)
Supplier: DIVERSIFIED BIOTECH
Description: Breathe-EASIER micropermeable membranes are ideal for cell growth, concentrating samples, and gas exchange. Formatted with an easy to use finger tab, ...
UOM: 1 * 100 items
Catalog Number: (KING756030)
Supplier: KING DISPOSABLES
Description: Sealing film, Length: 300 m, Width: 30 cm, PVC
UOM: 1 * 6 items
Supplier: KING DISPOSABLES
Description: Food wrap cling film of eight micron thickness.
Catalog Number: (PROOVHG-FKP30-R15)
Supplier: LGC Standards PROMOCHEM
Description: XRF thin film of 7,6 µm thickness and 76,2×15240 mm roll.
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 13 of 13
no targeter for Bottom