You Searched For: Microplate Seals
139  results were found
SearchResultCount:"139"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: Aluminium foils, 75,7 µm thick, for use with 96 or 384 well plates. Fit inside the rim of raised rim plates. These foils have one partial width end ta...
Supplier: VWR Collection
Description: These soft, non permeable 63,5 µm thick aluminium foils, with strong medical grade adhesive, eliminate the need for heat sealing devices or mats durin...
Supplier: VWR Collection
Description: VWR® WebSeal™, Microplate seal, Mat (96 wells), Single sheet/mat/strip, Sealmat (block cover), 96 square well shape, Well top shape: Square, Silicone, Clear, Non sterile
Supplier: VWR Collection
Description: Polyester film with acrylic adhesive. These non pierceable films in sheeted-roll format are recommended for ELISA tests, general incubation and short-...
Supplier: VWR Collection
Description: Identical to polyester films above, but include a colour dot and white writing area on each end tab for plate identification. End tabs with backing re...
Catalog Number: (391-0638)
Supplier: VWR Collection
Description: Easier to apply and remove as compared to standard moulded PCR strip caps.
UOM: 1 * 400 items
Supplier: VWR Collection
Description: Polyester seals feature a strong adhesive to minimize evaporation during PCR and storage applications.
Supplier: VWR Collection
Description: Aluminium foils with biologically inert adhesive. These foils are easily pierced by manual or automated pipette tips, for sample recovery without gene...
Supplier: VWR Collection
Description: VWR® MicroMat CLR, Microplate seal, Mat (96 wells), Single sheet/mat/strip, MicroMat CLR, Sealmat (block cover), 96 round well shape with domed base, Ø 7,0 mm, Well top shape: Round, EVA, Clear, Non sterile
Sale
Supplier: VWR Collection
Description: These heat resistant 74,9 µm thick films are designed for thermal cycling applications. Polypropylene films are not pierceable. For PCR applications, ...
Supplier: VWR Peqlab
Description: Easy to use adhesive films for PCR and qPCR plates.
Supplier: VWR Collection
Description: Adhesive sealing film or foil rolls constructed on plastic cores, for use with high throughput automated microplate sealers.
Catalog Number: (211-0350)
Supplier: VWR Collection
Description: VWR®, Microplate seal, Cap strip, Single sheet/mat/strip, Optically clear 8-cap strips for Real-Time PCR plates, PP
UOM: 1 * 125 items
Low Manufacturing Impact
Sustainable Packaging
Supplier: VWR Collection
Description: These 77,5 µm thick polyester films provide secure sealing around every well, not just a cover, thus minimising evaporation, preventing spillage and w...
Supplier: VWR Collection
Description: Transparent polyester films with strong, non absorbing, non fluorescing, medical grade adhesive for superior performance in qPCR applications. Supplie...
Sale
Supplier: VWR Collection
Description: 1x8 adhesive sealing strips are ideal for use with 1x8 strip-well ELISA plates.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 139
no targeter for Bottom