You Searched For: Scissors
110  results were found
SearchResultCount:"110"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: RSG
Description: Stainless steel, INOX 4021.
Supplier: RSG
Description: Stainless steel.
Supplier: RSG
Description: Carbon steel, nickel plated, 8/10 mm.
Supplier: RSG
Description: Stainless steel, with black plastic handles.
Catalog Number: (RSGA109.185)
Supplier: RSG
Description: Universal scissors with black plastic grip.
UOM: 1 * 1 items
Supplier: RSG
Description: Ice-tempered scissors with stainless steel blades and plastic covered handle.
Supplier: RSG
Description: Stainless steel.
Catalog Number: (233-1422)
Supplier: RSG
Description: Stainless steel, impact resistant plastic grip.
UOM: 1 * 1 items
Catalog Number: (233-1421)
Supplier: RSG
Description: Carbon steel.
UOM: 1 * 1 items
Catalog Number: (BOCH4151)
Supplier: BOCHEM
Description: Stainless steel bandage scissors with plastic handle.
UOM: 1 * 1 items
Catalog Number: (233-0222)
Supplier: OUTILS RUBIS SA
Description: Safety scissors with rounded tips.
UOM: 1 * 1 items
Catalog Number: (233-0218)
Supplier: OUTILS RUBIS SA
Description: Long legged multi-purpose scissors with rounded-off tips.
UOM: 1 * 1 items
Supplier: BOCHEM
Description: Straight paper scissors made of stainless steel.
Catalog Number: (BOCH4252)
Supplier: BOCHEM
Description: Scissors with stainless steel blades and red/black PP handle. Cuts tubes until 10 mm.
UOM: 1 * 1 items
Supplier: BOCHEM
Description: Straight scissors made of stainless steel.
Supplier: BOCHEM
Description: Stainless steel. Multi-purpose scissors for a wide range of applications.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 110
no targeter for Bottom