You Searched For: Retort Stands
88  results were found
SearchResultCount:"88"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (241-0089)
Supplier: VWR Collection
Description: Support plate, square.
UOM: 1 * 1 items
Supplier: VWR Collection
Description: The use of levelling platforms in electrophoresis can be a big help in maintaining a uniform gel thickness and therefore a uniform and quality sample ...
Supplier: VWR Collection
Description: Tripod stands (A): Cast iron tripod stand, painted with hammer finish, steel support rod, nickel plated.
Catalog Number: (241-0046)
Supplier: VWR Collection
Description: PU coated cast iron stand 200×450 mm, steel rod 500×12 mm, bosshead and holder for two funnels.
UOM: 1 * 1 items
Supplier: VWR Collection
Description: Roller bar platform, Roller bar platform, which has 4 adjustable cushioned rollers to hold glassware securely, 457×457 mm, bar size 457 mm, For: Standard/Advanced 3500 shakers
Supplier: VWR Collection
Description: H-shaped stands with nickel-coated steel rod and 420×400 mm grey marbled cast iron baseplate.
Supplier: VWR Collection
Description: PP, white.
Catalog Number: (444-0918)
Supplier: VWR Collection
Description: Cast iron support stand bases are constructed with black enamel finish for chemical resistance and durability.
UOM: 1 * 1 items
Catalog Number: (241-0045)
Supplier: VWR Collection
Description: PU coated, cast iron stand (200×450 mm), steel rod (500×12 mm), bosshead and holder for four funnels.
UOM: 1 * 1 items
Supplier: VWR Collection
Description: Mounting brackets for Labomeca® assemblies.
Catalog Number: (241-0104)
Supplier: VWR Collection
Description: PU coated cast iron stand, high grade steel rod 500×12 mm.
UOM: 1 * 1 items
Supplier: USBECK Laborgeräte
Description: Steel, powder coated.
Supplier: USBECK Laborgeräte
Description: Stainless steel. Can be used for a wide range of equipment, e.g. stirrers, hotplates etc.
Supplier: SCHOTT AG Lighting and imaging
Description: Brackets, EasyLED Spotlight "Plus" column adapter for Zeiss stand K, stand M
Supplier: USBECK Laborgeräte
Description: Powder-coated steel, thickness 10 mm.
Supplier: USBECK Laborgeräte
Description: Steel, powder coated. Firm, non slip stand. For a wide variety of uses. With M10 threaded hole on face side.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 88
no targeter for Bottom