You Searched For: Reaction Vessels
34  results were found
SearchResultCount:"34"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: Lenz Laborglas GmbH & CO.KG
Description: Reaction vessel, made of DURAN® borosilicate glass, round bottom, with easily readable graduation and laboratory flat flange (LF) with groove.
Supplier: Lenz Laborglas GmbH & CO.KG
Description: DURAN® borosilicate glass.
Supplier: Lenz Laborglas GmbH & CO.KG
Description: DURAN®, borosilicate glass, clear.
Catalog Number: (BOHLB320-01)
Supplier: Bohlender
Description: PFA, translucent.
UOM: 1 * 1 items
Supplier: Lenz Laborglas GmbH & CO.KG
Description: DURAN® borosilicate glass.
Supplier: Lenz Laborglas GmbH & CO.KG
Description: DURAN® borosilicate glass.
Supplier: Lenz Laborglas GmbH & CO.KG
Description: DURAN®, borosilicate glass.
Supplier: Lenz Laborglas GmbH & CO.KG
Description: Flask, round bottom, DURAN®, borosilicate glass, with flat flange with groove for vacuum use.
Supplier: Lenz Laborglas GmbH & CO.KG
Description: Reaction vessel, made of DURAN® borosilicate glass, flat bottom, with easily readable graduation and flat laboratory flange (DN) with groove. With two...
Supplier: Lenz Laborglas GmbH & CO.KG
Description: DURAN®, borosilicate glass, clear, with flat bottom.
Supplier: Lenz Laborglas GmbH & CO.KG
Description: DURAN®, borosilicate glass, clear, with round bottom.
Supplier: Bohlender
Description: PTFE, white.
Supplier: Bohlender
Description: PFA, translucent.
Supplier: DWK Life Sciences
Description: DURAN® borosilicate glass 3.3, clear, with laboratory flat flange (DN) with groove for vacuum use. Maximum operating pressure at 250 °C: 0,5 bar.
Supplier: WTW
Description: Flow vessels for WTW conductivity measuring cells are perfectly designed to perform measurements without the interference of carbon dioxide through th...
Catalog Number: (646-0035)
Supplier: DWK Life Sciences
Description: DURAN®, borosilicate glass 3.3, clear.
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 34
no targeter for Bottom