Beta storage racks for microtubes
113-0903EA 47.7 EUR
FLEM222093004 FLEM222093002 FLEM222093000
Beta storage racks for microtubes
Acrylic, crystal clear, thickness: 10 mm.
  • Stable and easy to clean
  • Accept 0,5 ml, 1,5 ml and 2 ml microtubes
Storage racks for use with 165×85×55 mm beta or gamma storage boxes (113-8830). Racks can hold up to 32 micro vials.