VWR®, Beta Radiation Bin
Supplier: VWR
VWR®
113-8993EA 715 EUR
113-8993
VWR®, Beta Radiation Bin
Boxes Beta/Gamma Radiation Boxes
Acrylic, transparent, thickness: 10 mm.
  • Attached hinged lid for easy filling and quick emptying
  • Robust, manoeuvrable and easy to clean
Maximum protection for short-term storage or disposal of mildly-contaminated materials.
W×D×H (inner): 285×285×580 mm.