You Searched For: Staining Racks
13  results were found
SearchResultCount:"13"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: GLASWARENFABRIK KARL HECHT
Description: Staining racks, made of Plexiglas®, with staining glass jars.
Catalog Number: (HECH41240010)
Supplier: GLASWARENFABRIK KARL HECHT
Description: Racks, made of Plexiglas®, for microscopic slides, with numbering 1 to 12 and field for inscriptions.
UOM: 1 * 1 items
Supplier: GLASWARENFABRIK KARL HECHT
Description: These staining racks are made of hardwood and have amber or clear glass pipette bottles.
Supplier: GLASWARENFABRIK KARL HECHT
Description: Staining rack, Plexiglas®, in the form of an extendable bridge, that can be fixed in the horizontal or at an inclined position. Suitable for trays or ...
Supplier: GLASWARENFABRIK KARL HECHT
Description: Racks that can be simply be added to a plain glass dish 220×150 mm.
Catalog Number: (631-0340)
Supplier: GLASWARENFABRIK KARL HECHT
Description: Stainless steel
UOM: 1 * 1 items
Supplier: DWK Life Sciences
Description: Soda-lime glass and stainless steel.
Supplier: DELTALAB
Description: Staining baskets and racks, made of stainless steel. Designed for 76×26 mm slides.
Catalog Number: (631-9520)
Supplier: KARTELL
Description: PP.
UOM: 1 * 1 items
Supplier: DELTALAB
Description: Stainless steel staining racks for microscope slides with and without dividers.
Supplier: GLASWARENFABRIK KARL HECHT
Description: Staining stands made of hardwood with staining glass jars.
Catalog Number: (VWRU15153-821)
Supplier: VWR Collection
Description: Stainless steel.
UOM: 1 * 1 items
Catalog Number: (720-0096)
Supplier: DIXON SCIENTIFIC
Description: Comprises two brass end-plates fitted with levelling screws and clips.
UOM: 1 * 2 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 13 of 13
no targeter for Bottom