You Searched For: Chromatography Vial Racks
15  results were found
SearchResultCount:"15"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: Vial Racks are ideal for storage and transportation of vials.
Catalog Number: (548-1388A)
Supplier: VWR Collection
Description: Rack, Chromatography vials , VWR®, PP, No. of holes: 50, W×D×H: 200×105×17 mm
UOM: 1 * 5 items
Catalog Number: (SPET268-816000)
Supplier: Thermo Fisher Scientific
Description: Sample rack holds up to 60 standard 14 ml vials.
UOM: 1 * 1 items
Supplier: PerkinElmer
Description: Rack, Chromatography vial, For tubes: 205  ml, Hole Ø: 61 mm, No. of holes: 10 (2×5)
Supplier: PerkinElmer
Description: These polypropylene racks accommodate 40 headspace auto-sampler vials.
Supplier: CHROMACOL
Description: These vial racks are chemically resistant and fairly robust for a safe working position on the lab bench and during transport.
Supplier: DWK Life Sciences
Description: These polypropylene racks accommodate 36 or 50 headspace auto-sampler vials. Each well has an opening in the bottom to facilitate drainage.
Supplier: DWK Life Sciences
Description: These PP racks accommodate 48, 50, 90 or 96 vials of different sizes.
Supplier: MACHEREY-NAGEL
Description: These chromatography vial racks are used for a safe standing position of the vials on the lab bench.
Supplier: Restek
Description: These vial racks securely hold up to 36 headspace vials.
Catalog Number: (HEWL5067-0243)
Supplier: Agilent
Description: Stackable storage racks holds up to 50 standard 2 ml, 12 mm vials.
UOM: 1 * 1 items
Supplier: NEOLAB
Description: PC, transparent.
Supplier: Agilent
Description: Rack, Chromatography vial
Supplier: Merck
Description: PP, red.
Supplier: NSC (NATIONAL SCIENTIFIC COMP.
Description: Stackable PP or PS storage racks for 12×32 mm or 15×45 mm vials.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 15 of 15
no targeter for Bottom