You Searched For: Racks
475  results were found
SearchResultCount:"475"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: Accessory for spectrophotometers, Cuvette, VWR®
Supplier: VWR Collection
Description: Freezer racks as storage and inventory systems for upright ULT freezers makes it easier organise your samples.
Sale
Supplier: VWR Collection
Description: These test tube racks are either made from rust-proof, electrolytically polished stainless steel or from polyamide coated steel.
Sale
Supplier: VWR Collection
Description: Nylon, glass fibre reinforced.
Sale
Supplier: VWR Collection
Description: Racks, Test tube, VWR®, Stainless steel test tube rack for 12 and 18 l baths, No. of holes: 119, For tubes: 0,5 ml
Supplier: VWR Collection
Description: PP
Supplier: VWR Collection
Description: Freezer racks as storage and inventory systems for upright ULT freezers makes it easier organise your samples.
Sale
Supplier: VWR Collection
Description: PP.
Supplier: VWR Collection
Description: Tube racks suitable for: VWB 6, VWB 12, VWB 18, VWB 26, maximum 1, 4, 7, 7 racks per bath respectively.
Supplier: VWR Collection
Description: Freezer racks as storage and inventory systems for upright ULT freezers makes it easier organise your samples.
Sale
Catalog Number: (525-0972)
Supplier: VWR Collection
Description: Rack, PP, white. This 'open style' floating rack provides tubes with maximum contact with the water and better circulation of the water around the tub...
UOM: 1 * 2 items
Supplier: VWR Collection
Description: PP.
Supplier: VWR Collection
Description: Rack, Test tube, VWR®, Stainless steel test tube rack, Stainless steel, For tube Ø: 10 mm
Supplier: VWR Collection
Description: PP rack with four sides that can be loaded independently.
Supplier: VWR Collection
Description: Freezer racks as storage and inventory systems for upright ULT freezers makes it easier organise your samples.
Supplier: VWR Collection
Description: PP.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 475
no targeter for Bottom